Glycine methyl ester hydrochloride (H-Gly-OMe.HCl) 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl glycinate hydrochloride, H-Gly-OMe.HCl, Glycine Methyl Ester HCl

A14083-0E A14083-22 A14083-36
CAAAA14083-0EEA 764.72 CAD
CAAAA14083-0E CAAAA14083-22 CAAAA14083-36
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl) 99%
Glycine methyl ester hydrochloride (H-Gly-OMe.HCl)

Purity: 99% CAS No.: 5680-79-5 Molecular Formula: H2NCH2CO2CH3.HCl Formula Weight: 125.56 Melting Point: ca 175° dec. Beilstein Reference: 3593644

Formula: C₃H₈ClNO₂
Melting Pt: ca 175 °C (dec.)
Density: 1.000
Storage Temperature: Ambient
MDL Number: MFCD00012870
CAS Number: 5680-79-5
EINECS: 227-139-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR