4-Hydroxy-2(5H)-furanone 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Tetronic acid, beta-Tetronic acid, 2,4(3H,5H)-Furandione, Tetrahydrofuran-2,4-dione
A14079-06
A14079-14
CAAAA14079-06EA
660.44
CAD
CAAAA14079-06
CAAAA14079-14
4-Hydroxy-2(5H)-furanone 96%
4-Hydroxy-2(5H)-furanone (Tetrahydrofuran-2,4-dione, Tetronic acid, β-Oxo-γ-butyrolactone)
Purity: 96%
CAS No.: 541-57-1
Molecular Formula: C4H4O3
Formula Weight: 100.07
Melting Point: ca 140° dec.
Beilstein Reference: 108442
Formula:
C₄H₄O₃ Melting Pt: ca 140 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00010086 CAS Number: 541-57-1 EINECS: 208-785-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...