Diethyl acetamidomalonate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diethyl 2-acetamidomalonate

A14048-14 A14048-22 A14048-36
CAAAA14048-14EA 38.71 CAD
CAAAA14048-14 CAAAA14048-22 CAAAA14048-36
Diethyl acetamidomalonate 98+%
Diethyl acetamidomalonate

Purity: 98+% CAS No.: 1068-90-2 Molecular Formula: CH3CONHCH(CO2CH2CH3)2 Formula Weight: 217.22 Melting Point: 96-99° Boiling Point: 185°/20mm MDL No.: MFCD00009146

Formula: C₉H₁₅NO₅
MW: 217.22 g/mol
Boiling Pt: 185 °C/20 mm
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00009146
CAS Number: 1068-90-2
EINECS: 213-952-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR