tert-Butyl hydroperoxide ≥70% in aqueous solution
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13926-0F
A13926-AE
A13926-AP
CAAAA13926-0FEA
453.46
CAD
CAAAA13926-0F
CAAAA13926-AE
CAAAA13926-AP
tert-Butyl hydroperoxide ≥70% in aqueous solution
tert-Butyl hydroperoxide
Fieser: 1,88 13,51 14,60 15,55 16,53 17,56 18,71 19,51 20,61 21,76
CAS No.: 75-91-2
Molecular Formula: C4H10O2
Formula Weight: 90.12
Storage Temperature: 2°C to 8°C
Flash Point: 43°(109°F)
Physical Form: Liquid
Density: 0.937
Refractive Index: 1.3870
Appearance: Colorless
Melting Point: -3°
Boiling Point: 96° dec.
Beilstein Reference: 1098280
Merck Reference: 14,1570
UN No.: 3109
MDL No.: MFCD00002130
Formula:
C₄H₁₀O₂ Boiling Pt: 96 °C (dec.) Melting Pt: –3 °C Density: 0.937 Flash Pt: 43 °C(109 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00002130 CAS Number: 75-91-2 EINECS: 200-915-7 UN: 3109 ADR: 5.2, Merck Index: 14,1570 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...