3-Sulfolene 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-sulfolene, 3-sulpholene, Butadiene sulfone, Butadiene sulphone

A13887-0B A13887-0I A13887-22 A13887-36
CAAAAA13887-0BEA 170.64 CAD
CAAAAA13887-0B CAAAAA13887-0I CAAAAA13887-22 CAAAAA13887-36
3-Sulfolene 98%
3-Sulpholene

Fieser: 2,389 3,272 4,468 12,455


Purity: 98% CAS No.: 77-79-2 Molecular Formula: C4H6O2S Formula Weight: 118.15 Flash Point: 112°(233°F) Melting Point: 63-67° Beilstein Reference: 107004 Merck Reference: 14,8956 MDL No.: MFCD00005481

Formula: C₄H₆O₂S
MW: 118.16 g/mol
Melting Pt: 63…67 °C
Density: 1.314
Flash Pt: 112 °C(233 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005481
CAS Number: 77-79-2
EINECS: 201-059-7
UN: 3335
ADR: "9",
Merck Index: 14,8956

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR