Benzylphosphonic acid 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13850-03
A13850-06
A13850-14
CAAAA13850-03EA
51.51
CAD
CAAAA13850-03
CAAAA13850-06
CAAAA13850-14
Benzylphosphonic acid 97%
Benzylphosphonic acid
Structural mimic of phosphotyrosine.
Purity: 97%
CAS No.: 6881-57-8
Molecular Formula: C6H5CH2PO3H2
Formula Weight: 172.11
Melting Point: 166-172°
Beilstein Reference: 2519318
UN No.: 3261
MDL No.: MFCD00039519
Formula:
C₇H₉O₃P Melting Pt: 166…172 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039519 CAS Number: 6881-57-8 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...