N-(Hydroxymethyl)acetamide 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
N-Hydroxymethylacetamide, N-Acetylmethanolamine, Acetamidomethanol
A13820-09
A13820-14
CAAAAA13820-09EA
95.59
CAD
CAAAAA13820-09
CAAAAA13820-14
N-(Hydroxymethyl)acetamide 98%
N-(Hydroxymethyl)acetamide
Fieser: 2,4
Purity: 98%
CAS No.: 625-51-4
Molecular Formula: CH3CONHCH2OH
Formula Weight: 89.09
Storage Temperature: 2°C to 8°C
Melting Point: 47-52°
Beilstein Reference: 506226
Merck Reference: 14,4242
MDL No.: MFCD00014417
Formula:
C₃H₇NO₂ MW: 89.09 g/mol Melting Pt: 47…52 °C Density: 1.14 Storage Temperature: Refrigerator |
MDL Number:
MFCD00014417 CAS Number: 625-51-4 EINECS: 210-897-2 Merck Index: 14,4242 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...