Glycylglycylglycine (Gly-Gly-Gly)
Supplier: Thermo Scientific Chemicals
A13778-03
A13778-06
A13778-14
CAAAA13778-03EA
41.71
CAD
CAAAA13778-03
CAAAA13778-06
CAAAA13778-14
Glycylglycylglycine (Gly-Gly-Gly)
Proteins and Peptides
- EINECS: 209-122-0
- Melting point: ∼240 °C (dec.)
- Merck index_V14: 14,6579
Purity: 98%
CAS No.: 556-33-2
Molecular Formula: H2NCH2(CONHCH2)2CO2H
Formula Weight: 191.14
Melting Point: ca 240° dec.
Beilstein Reference: 1711130
Merck Reference: 14,6579
MDL No.: MFCD00036223
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...