3-(4-Fluorobenzoyl)propionic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A13755-06
A13755-14
CAAAA13755-06EA
44.87
CAD
CAAAA13755-06
CAAAA13755-14
3-(4-Fluorobenzoyl)propionic acid 97%
3-(4-Fluorobenzoyl)propionic acid
Purity: 97%
CAS No.: 366-77-8
Molecular Formula: C10H9FO3
Formula Weight: 196.18
Odor: Odorless
Melting Point: 99-103°
Beilstein Reference: 958145
MDL No.: MFCD00002793
Formula:
C₁₀H₉FO₃ MW: 196.18 g/mol Melting Pt: 99…103 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002793 CAS Number: 366-77-8 EINECS: 206-679-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...