EPPS (HEPPS, 4-(2-Hydroxyethyl)-1-piperazinepropanesulfonic acid) 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-[4-(2-Hydroxyethyl)-1-piperazinyl]propanesulfonic acid, 4-(2-Hydroxyethyl)-1-piperazinepropanesulphonic acid, N-(2-Hydroxyethyl)piperazine-N′-(3-propanesulfonic acid), N-(2-Hydroxyethyl)piperazine-N′-(3-propanesulphonic acid), HEPPS, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]-propanesulfonic acid, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]propanesulphonic acid, 4-(2-Hydroxyethyl)piperazine-1-propanesulfonic acid, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]-propanesulphonic acid, 4-(2-Hydroxyethyl)-1-piperazinepropanesulfonic acid, 4-(2-Hydroxyethyl)piperazine-1-propanesulphonic acid
Purity: 99%
CAS No.: 16052-06-5
Molecular Formula: C9H20N2O4S
Formula Weight: 252.33
Melting Point: 242° dec.
Beilstein Reference: 3957385
Formula:
C₉H₂₀N₂O₄S MW: 252.33 g/mol Melting Pt: 242 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006160 CAS Number: 16052-06-5 EINECS: 240-198-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...