2-Chloro-N,N-diethylacetamide 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A13691-14
A13691-22
CAAAA13691-14EA
129.24
CAD
CAAAA13691-14
CAAAA13691-22
2-Chloro-N,N-diethylacetamide 97%
2-Chloro-N,N-diethylacetamide
Purity: 97%
CAS No.: 2315-36-8
Molecular Formula: C6H12ClNO
Formula Weight: 149.62
Flash Point: 113°(235°F)
Refractive Index: 1.4700
Boiling Point: 123-124°/15mm
Beilstein Reference: 605866
UN No.: 2810
MDL No.: MFCD00000928
Formula:
C₆H₁₂ClNO Boiling Pt: 123…124 °C/15 mm Density: 1.087 Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000928 CAS Number: 2315-36-8 EINECS: 219-019-2 UN: 2810 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...