Ethylenediammonium dichloride 98%

Supplier: Thermo Scientific Chemicals
Warning

A13563-0E A13563-22 A13563-36
CAAAA13563-0EEA 564.06 CAD
CAAAA13563-0E CAAAA13563-22 CAAAA13563-36
Ethylenediammonium dichloride 98%
Ethylenediammonium dichloride

Purity: 98% CAS No.: 333-18-6 Molecular Formula: HCl.H2NCH2CH2NH2.HCl Formula Weight: 133.02 Melting Point: 300° MDL No.: MFCD00012524

Formula: C₂H₁₀Cl₂N₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00012524
CAS Number: 333-18-6
EINECS: 206-369-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR