Benzylideneacetone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Phenylbut-3-en-2-one, 4-Phenyl-3-buten-2-one
A13537-0B
A13537-30
CAAAA13537-0BEA
153.73
CAD
CAAAA13537-0B
CAAAA13537-30
Benzylideneacetone 98+%
Benzylideneacetone
Purity: 98+%
CAS No.: 122-57-6
Molecular Formula: C10H10O
Formula Weight: 146.19
Flash Point: 116°(241°F)
Physical Form: Crystalline
Density: 1.038
Sensitivity: Light Sensitive
Odor: Characteristic
Melting Point: 38-42°
Boiling Point: 261°
Beilstein Reference: 742046
Merck Reference: 14,1137
MDL No.: MFCD00008779
Formula:
C₁₀H₁₀O MW: 146.19 g/mol Boiling Pt: 261 °C Melting Pt: 38…42 °C Density: 1.038 Flash Pt: 116 °C (241 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008779 CAS Number: 122-57-6 EINECS: 204-555-1 Merck Index: 14,1137 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...