L(+)-Citrulline 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Cit-OH
A13316-09
A13316-18
CAAAA13316-09EA
82.48
CAD
CAAAA13316-09
CAAAA13316-18
L(+)-Citrulline 98%
L(+)-Citrulline
Purity: 98%
CAS No.: 372-75-8
Molecular Formula: C6H13N3O3
Formula Weight: 175.19
Melting Point: 212-214° dec.
Beilstein Reference: 6055157
Merck Reference: 14,2331
Optical Rotation: +22° (c=2 in 1N HCl)
MDL No.: MFCD00064397
Formula:
C₆H₁₃N₃O₃ MW: 175.19 g/mol Melting Pt: 212…214 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064397 CAS Number: 372-75-8 EINECS: 206-759-6 Merck Index: 14,2331 |
Specification Test Results
Optical rotation | +22° (c=2 in 1N HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...