Sarcosine ethyl ester hydrochloride 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: Sarcosine Ethyl Ester HCl, H-Sar-OEt·HCl, Ethyl 2-(methylamino)acetate hydrochloride

A13112-14 A13112-22
CAAAA13112-14EA 139.2 CAD
CAAAA13112-14 CAAAA13112-22 CAAAA13112-36
Sarcosine ethyl ester hydrochloride 98+%
Sarcosine ethyl ester hydrochloride

Purity: 98+% CAS No.: 52605-49-9 Molecular Formula: C5H11NO2.HCl Formula Weight: 153.61 Storage Temperature: 2°C to 8°C Melting Point: 122-126° Beilstein Reference: 3911182 MDL No.: MFCD00012506

Formula: C₅H₁₁NO₂·HCl
MW: 153.61
Melting Pt: 122…126 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012506
CAS Number: 52605-49-9
EINECS: 258-037-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR