5-Decyne 98%

Supplier: Thermo Scientific Chemicals
Warning

A13050-09 A13050-18
CAAAA13050-09EA 105.39 CAD
CAAAA13050-09 CAAAA13050-18
5-Decyne 98%
5-Decyne

Purity: 98% CAS No.: 1942-46-7 Molecular Formula: CH3(CH2)3C≡C(CH2)3CH3 Formula Weight: 138.26 Flash Point: 54°(129°F) Refractive Index: 1.4330 Odor: Petrol-like Boiling Point: 177-178° Beilstein Reference: 1697840 UN No.: 3295 MDL No.: MFCD00027277

Formula: C₁₀H₁₈
MW: 138.25 g/mol
Boiling Pt: 177…178 °C
Density: 0.770
Flash Pt: 54 °C(129 °F)
Storage Temperature: Ambient
MDL Number: MFCD00027277
CAS Number: 1942-46-7
UN: 3295
ADR: "3",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR