N-Acetyl-β-D-glucosamine 98+%

Supplier: Thermo Scientific Chemicals

A13047-22 A13047-09 A13047-18
CAAAAA13047-22EA 216.46 CAD
CAAAAA13047-22 CAAAA13047-09 CAAAA13047-18
N-Acetyl-β-D-glucosamine 98+%
N-Acetyl-β-D-glucosamine

Purity: 98+% CAS No.: 7512-17-6 Molecular Formula: C8H15NO6 Formula Weight: 221.21 Storage Temperature: 2°C to 8°C Melting Point: 213° dec. Beilstein Reference: 1727589 Merck Reference: 14,4458 Optical Rotation: +42° (c=2 in water) MDL No.: MFCD00136044

Formula: C₈H₁₅NO₆
Melting Pt: 213 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00136044
CAS Number: 7512-17-6
EINECS: 231-368-2
Merck Index: 14,4458

Order Now

Specification Test Results

Optical rotation +42° (c=2 in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR