Benzyl carbazate 98+%

Supplier: Thermo Scientific Chemicals

A13005-06 A13005-14 A13005-22
CAAAAA13005-06EA 70.78 CAD
CAAAAA13005-06 CAAAAA13005-14 CAAAAA13005-22
Benzyl carbazate 98+%
Benzyl carbazate

Fieser: 1,110


Purity: 98+% CAS No.: 5331-43-1 Molecular Formula: C8H10N2O2 Formula Weight: 166.18 Melting Point: 66-70° Beilstein Reference: 1952982

Formula: C₈H₁₀N₂O₂
Melting Pt: 66…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00041890
CAS Number: 5331-43-1
EINECS: 226-230-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR