6-Chloro-1,3-dimethyluracil 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12755-03
A12755-06
CAAAA12755-03EA
24.65
CAD
CAAAA12755-03
CAAAA12755-06
6-Chloro-1,3-dimethyluracil 97%
6-Chloro-1,3-dimethyluracil
Purity: 97%
CAS No.: 6972-27-6
Molecular Formula: C6H7ClN2O2
Formula Weight: 174.59
Storage Temperature: -30°C to -10°C
Melting Point: 109-111°
Beilstein Reference: 144393
MDL No.: MFCD00038066
Formula:
C₆H₇ClN₂O₂ Melting Pt: 109…111 °C Storage Temperature: Freezer |
MDL Number:
MFCD00038066 CAS Number: 6972-27-6 EINECS: 230-205-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...