Silicone oil for oil baths –40…+200°C

Supplier: Thermo Scientific Chemicals

A12728-0E A12728-22 A12728-36
CAAAA12728-0EEA 507.18 CAD
CAAAA12728-0E CAAAA12728-22 CAAAA12728-36
Silicone oil for oil baths –40…+200°C
Silicone oil

Silicone oil is used as a calibration media in temperature controlled baths in laboratories due to its good thermal characteristics and low flammability.


It is also used as active component in silicone grease, lubricants and heat transfer fluids. It finds application in cosmetics, hair conditioners and as a sorbent for the analysis of headspace (dissolved gas analysis) of food.


Viscosity ~ 40 cSt at 20 °C.


Caution: Incompatible with strong oxidizing agents.

Density: 0.963
Flash Pt: >270 °C(518 °F)
Storage Temperature: Ambient
MDL Number: MFCD00132673
CAS Number: 68083-14-7
Merck Index: 14,8495

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR