Ethylene chlorophosphite 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12724-14
A12724-22
A12724-36
CAAAA12724-14EA
290.72
CAD
CAAAA12724-14
CAAAA12724-22
CAAAA12724-36
Ethylene chlorophosphite 97%
Ethylene chlorophosphite
Fieser: 1,372
Purity: 97%
CAS No.: 822-39-9
Molecular Formula: C2H4ClO2P
Formula Weight: 126.47
Storage Temperature: 2°C to 8°C
Flash Point: 86°(186°F)
Refractive Index: 1.4910
Boiling Point: 46-47°/15mm
Beilstein Reference: 506179
UN No.: 3265
MDL No.: MFCD00014511
Formula:
C₂H₄ClO₂P Boiling Pt: 46…47 °C/15 mm Density: 1.415 Flash Pt: 86 °C(186 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00014511 CAS Number: 822-39-9 EINECS: 212-499-4 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...