4,4-Dimethyl-2-oxazoline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12670-22
A12670-06
A12670-14
CAAAAA12670-22EA
1072.82
CAD
CAAAAA12670-22
CAAAA12670-06
CAAAA12670-14
4,4-Dimethyl-2-oxazoline 98%
4,4-Dimethyl-2-oxazoline
Purity: 98%
CAS No.: 30093-99-3
Molecular Formula: C5H9NO
Formula Weight: 99.13
Flash Point: 1°(33°F)
Refractive Index: 1.4190
Boiling Point: 99-100°
Beilstein Reference: 1209606
UN No.: 1993
MDL No.: MFCD00014507
Formula:
C₅H₉NO Boiling Pt: 99…100 °C Density: 0.940 Flash Pt: 1 °C(33 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014507 CAS Number: 30093-99-3 EINECS: 250-045-7 UN: 1993 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...