5-Methylbenzo[b]thiophene 97%

Supplier: Thermo Scientific Chemicals
Warning

A12666-03 A12666-06
CAAAA12666-03EA 170.64 CAD
CAAAA12666-03 CAAAA12666-06
5-Methylbenzo[b]thiophene 97%
5-Methylbenzo[b]thiophene

Purity: 97% CAS No.: 14315-14-1 Molecular Formula: C9H8S Formula Weight: 148.23 Melting Point: 32-36° Boiling Point: 71-73°/0.6mm MDL No.: MFCD00052509

Formula: C₉H₈S
Boiling Pt: 71…73 °C/0.6 mm
Melting Pt: 32…36 °C
Density: 1.11
Storage Temperature: Ambient
MDL Number: MFCD00052509
CAS Number: 14315-14-1
EINECS: 238-256-2
UN: 3335

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR