L(+)-Glutamic acid hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Danger

A12505-0B A12505-30
CAAAA12505-0BEA 126.08 CAD
CAAAA12505-0B CAAAA12505-30
L(+)-Glutamic acid hydrochloride 99%
L(+)-Glutamic acid hydrochloride

Purity: 99% CAS No.: 138-15-8 Molecular Formula: C5H9NO4.HCl Formula Weight: 183.59 Melting Point: 203° dec. Beilstein Reference: 3565569 Merck Reference: 14,4469 Optical Rotation: +23° (c=6 in water) MDL No.: MFCD00012619

Formula: C₅H₁₀ClNO₄
Melting Pt: 203 °C (dec.)
Density: 1.525
Storage Temperature: Ambient
MDL Number: MFCD00012619
CAS Number: 138-15-8
EINECS: 205-315-9
Merck Index: 14,4469

Order Now

Specification Test Results

Optical rotation +23° (c=6 in water)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR