Benzo[b]thiophene-2-carboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12471-14
A12471-03
A12471-06
CAAAAA12471-14EA
646.22
CAD
CAAAAA12471-14
CAAAA12471-03
CAAAA12471-06
Benzo[b]thiophene-2-carboxylic acid 98%
Benzo[b]thiophene-2-carboxylic acid
Purity: 98%
CAS No.: 6314-28-9
Molecular Formula: C9H6O2S
Formula Weight: 178.22
Melting Point: 236-242°
Beilstein Reference: 124613
Merck Reference: 14,9345
MDL No.: MFCD00051636
Formula:
C₉H₆O₂S Melting Pt: 236…242 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051636 CAS Number: 6314-28-9 Merck Index: 14,9345 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...