1,8-Diazabicyclo[5.4.0]undec-7-ene 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,3,4,6,7,8,9,10-Octahydropyrimido[1,2-a]azepine, DBU

A12449-14 A12449-22 A12449-36
CAAAA12449-14EA 61.46 CAD
CAAAA12449-14 CAAAA12449-22 CAAAA12449-36
1,8-Diazabicyclo[5.4.0]undec-7-ene 99%
1,8-Diazabicyclo[5.4.0]undec-7-ene

Fieser: 2,101 14,109 15,109 16,105 17,99 18,120 19,117 20,129 21,158


Purity: 99% CAS No.: 6674-22-2 Molecular Formula: C9H16N2 Formula Weight: 152.24 Flash Point: 116°(240°F) Refractive Index: 1.5220 Odor: Amine-like pH: 12.8 Melting Point: -70° Boiling Point: 261° Beilstein Reference: 508906 UN No.: 3267 MDL No.: MFCD00006930

Formula: C₉H₁₆N₂
Boiling Pt: 261 °C
Melting Pt: –70 °C
Density: 1.020
Flash Pt: 116 °C(240 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006930
CAS Number: 6674-22-2
EINECS: 229-713-7
UN: 2922
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR