Ethyl-3-bromopropionate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12402-22
A12402-36
CAAAA12402-22EA
293.88
CAD
CAAAA12402-22
CAAAA12402-36
Ethyl-3-bromopropionate 99%
Ethyl 3-bromopropionate
Purity: 99%
CAS No.: 539-74-2
Molecular Formula: BrCH2CH2CO2CH2CH3
Formula Weight: 181.03
Flash Point: 79°(174°F)
Refractive Index: 1.4520
Boiling Point: 67-71°/10mm
Beilstein Reference: 1700933
MDL No.: MFCD00000251
Formula:
C₅H₉BrO₂ Boiling Pt: 67…71 °C/10 mm Density: 1.416 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000251 CAS Number: 539-74-2 EINECS: 208-724-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...