S-Methyl thioacetate 98+%

Supplier: Thermo Scientific Chemicals
Danger

A12400-06 A12400-14 A12400-22
CAAAA12400-06EA 121.34 CAD
CAAAA12400-06 CAAAA12400-14 CAAAA12400-22
S-Methyl thioacetate 98+%
S-Methyl thioacetate

Purity: 98+% CAS No.: 1534-08-3 Molecular Formula: C3H6OS Formula Weight: 91.14 Flash Point: 12°(54°F) Odor: Unpleasant Boiling Point: 97-99° UN No.: 1993 MDL No.: MFCD00014989

Formula: C₃H₆OS
Boiling Pt: 97…99 °C
Density: 1.024
Flash Pt: 12 °C(54 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014989
CAS Number: 1534-08-3
EINECS: 216-252-1
UN: 1993
ADR: "3",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR