tert-Butyl carbazate 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12383-36
A12383-14
A12383-22
CAAAAA12383-36EA
938.52
CAD
CAAAAA12383-36
CAAAA12383-14
CAAAA12383-22
tert-Butyl carbazate 98+%
tert-Butyl carbazate
Fieser: 1,85 2,46 11,88
Purity: 98+%
CAS No.: 870-46-2
Molecular Formula: C5H12N2O2
Formula Weight: 132.16
Storage Temperature: 2°C to 8°C
Flash Point: 91°(195°F)
Melting Point: 37-41°
Boiling Point: 63-65°/0.1mm
Beilstein Reference: 1756967
UN No.: 1325
MDL No.: MFCD00007593
Formula:
C₅H₁₂N₂O₂ Boiling Pt: 63…65 °C/0.1 mm Melting Pt: 37…41 °C Flash Pt: 91 °C(195 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00007593 CAS Number: 870-46-2 EINECS: 212-795-3 UN: 2926 ADR: "4.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...