Ethyl (ethoxymethylene)cyanoacetate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Ethyl 2-Cyano-3-Ethoxyacrylate
A12366-0B
A12366-18
A12366-30
CAAAA12366-0BEA
613.04
CAD
CAAAA12366-0B
CAAAA12366-18
CAAAA12366-30
Ethyl (ethoxymethylene)cyanoacetate 98%
Ethyl (ethoxymethylene)cyanoacetate
Fieser: 1,380
Purity: 98%
CAS No.: 94-05-3
Molecular Formula: C8H11NO3
Formula Weight: 169.18
Flash Point: 130°(266°F)
Melting Point: 47-52°
Boiling Point: 190-191°/30mm
Beilstein Reference: 878814
UN No.: 3439
MDL No.: MFCD00009136
Formula:
C₈H₁₁NO₃ MW: 169.18 g/mol Boiling Pt: 190…191 °C/30 mm Melting Pt: 47…52 °C Flash Pt: 130 °C(266 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009136 CAS Number: 94-05-3 EINECS: 202-299-5 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...