2,2'-Bithiophene 98%

Supplier: Thermo Scientific Chemicals
Warning

A12335-14 A12335-03 A12335-06
CAAAAA12335-14EA 467.68 CAD
CAAAAA12335-14 CAAAA12335-03 CAAAA12335-06
2,2'-Bithiophene 98%
2,2'-Bithiophene

Purity: 98% CAS No.: 492-97-7 Molecular Formula: C8H6S2 Formula Weight: 166.27 Flash Point: 110°(230°F) Melting Point: 30-33° Boiling Point: 260° Beilstein Reference: 3039 MDL No.: MFCD00005414

Formula: C₈H₆S₂
Boiling Pt: 260 °C
Melting Pt: 30…33 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005414
CAS Number: 492-97-7
EINECS: 207-767-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR