Pyridoxal-5'-phosphate monohydrate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Pyridoxal-5-phosphate hydrate, Pyridoxal 5-phosphate monohydrate, Pyridoxal 5'-phosphate monohydrate, (4-Formyl-5-hydroxy-6-methylpyridin-3-yl)methyl dihydrogen phosphate hydrate

A12323-03 A12323-06 A12323-14
CAAAA12323-03EA 68.57 CAD
CAAAA12323-03 CAAAA12323-06 CAAAA12323-14
Pyridoxal-5'-phosphate monohydrate 98%
Pyridoxal 5'-phosphate monohydrate

Active form of Vitamin B-6.


Purity: 98% CAS No.: 41468-25-1 Molecular Formula: C8H10NO6P.H2O Formula Weight: 265.17 (247.15anhy) Storage Temperature: -30°C to -10°C Melting Point: 142° MDL No.: MFCD00149414

Formula: C₈H₁₀NO₆P·1H₂O
MW: 265.17 g/mol (247.15 anhy)
Melting Pt: 142 °C
Storage Temperature: Freezer
MDL Number: MFCD00149414
CAS Number: 41468-25-1
EINECS: 200-208-3
Merck Index: 14,7979

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR