2,6-Dichloroaniline 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12320-22
A12320-36
CAAAA12320-22EA
120.55
CAD
CAAAA12320-22
CAAAA12320-36
2,6-Dichloroaniline 98%
2,6-Dichloroaniline
Purity: 98%
CAS No.: 608-31-1
Molecular Formula: C6H5Cl2N
Formula Weight: 162.02
Flash Point: 118°(244°F)
Melting Point: 37-40°
Boiling Point: 97°/5mm
Beilstein Reference: 775084
UN No.: 3442
MDL No.: MFCD00007675
Formula:
C₆H₅Cl₂N Boiling Pt: 97 °C/5 mm Melting Pt: 37…40 °C Density: 1.28 Flash Pt: 118 °C (244 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007675 CAS Number: 608-31-1 EINECS: 210-160-5 UN: 3442 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...