Phthalazine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzo[d]pyridazine

A12270-06 A12270-14
CAAAA12270-06EA 115.18 CAD
CAAAA12270-06 CAAAA12270-14
Phthalazine 98%
Phthalazine

Purity: 98% CAS No.: 253-52-1 Molecular Formula: C8H6N2 Formula Weight: 130.15 Melting Point: 89-92° Boiling Point: 317° Merck Reference: 14,7370 MDL No.: MFCD00006908

Formula: C₈H₆N₂
MW: 130.15 g/mol
Boiling Pt: 317 °C
Melting Pt: 89…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00006908
CAS Number: 253-52-1
EINECS: 205-963-2
Merck Index: 14,7370

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR