Calcium bis(dihydrogenorthophosphate) hydrate 97%

Supplier: Thermo Scientific Chemicals
Warning

A12123-0C A12123-0E A12123-36
CAAAAA12123-0CEA 873.74 CAD
CAAAAA12123-0C CAAAAA12123-0E CAAAAA12123-36
Calcium bis(dihydrogenorthophosphate) hydrate 97%
Calcium bis(dihydrogenorthophosphate) hydrate

Applications: pH control reagent, stabilizer for plastics, mfg. of glass
Loses H2O at 100°, dec. at 200°


Purity: 97% CAS No.: 301524-28-7 Molecular Formula: Ca(H2PO4)2.xH2O Formula Weight: 234.06(anhy) Flash Point: None Melting Point: ca 200° dec. MDL No.: MFCD00010898

Formula: H₄CaO₈P₂·H₂O
Melting Pt: ca 200 °C (dec.)
Flash Pt: None
Storage Temperature: Ambient
MDL Number: MFCD00010898
CAS Number: 301524-28-7
EINECS: 231-837-1
Merck Index: 14,1693

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR