L(+)-Ornithine hydrochloride 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Orn-OH Hydrochloride, H-Orn-OH·HCl
A12111-14
A12111-22
A12111-36
CAAAA12111-14EA
57.51
CAD
CAAAA12111-14
CAAAA12111-22
CAAAA12111-36
L(+)-Ornithine hydrochloride 99%
L(+)-Ornithine hydrochloride
Purity: 99%
CAS No.: 3184-13-2
Molecular Formula: C5H12N2O2.HCl
Formula Weight: 168.62
Melting Point: ca 238° dec.
Beilstein Reference: 3625847
Merck Reference: 14,6874
Optical Rotation: +23° (c=5 in 5N HCl)
MDL No.: MFCD00064562
Formula:
C₅H₁₃ClN₂O₂ Melting Pt: ca 238 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064562 CAS Number: 3184-13-2 EINECS: 221-678-6 Merck Index: 14,6874 |
Specification Test Results
Optical rotation | +23° (c=5 in 5N HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...