Diethyl methylmalonate 99%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Diethyl 2-methylmalonate
A12078-0B
A12078-18
A12078-30
CAAAA12078-0BEA
461.36
CAD
CAAAA12078-0B
CAAAA12078-18
CAAAA12078-30
Diethyl methylmalonate 99%
Diethyl methylmalonate
Fieser: 5,216
Purity: 99%
CAS No.: 609-08-5
Molecular Formula: C8H14O4
Formula Weight: 174.20
Flash Point: 85°(185°F)
Refractive Index: 1.4130
Boiling Point: 198-199°
Beilstein Reference: 637028
MDL No.: MFCD00009162
Formula:
C₈H₁₄O₄ Boiling Pt: 198…199 °C Density: 1.02 Flash Pt: 85 °C(185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009162 CAS Number: 609-08-5 EINECS: 210-175-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...