Isethionic acid sodium salt (sodium 2-hydroxyethanesulfonate) 98%

Supplier: Thermo Scientific Chemicals

A12054-0B A12054-0I
CAAAA12054-0BEA 109.65 CAD
CAAAA12054-0B CAAAA12054-0I
Isethionic acid sodium salt (sodium 2-hydroxyethanesulfonate) 98%
Isethionic acid sodium salt (Sodium 2-hydroxyethanesulphonate)

A surfactant and skin and mucous membrane irritant.


Purity: 98% CAS No.: 1562-00-1 Molecular Formula: HO(CH2)2SO3Na Formula Weight: 148.11 Melting Point: 191-194° Beilstein Reference: 3633992

Formula: C₂H₅NaO₄S
Melting Pt: 191…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00007534
CAS Number: 1562-00-1
EINECS: 216-343-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR