Methyl tiglate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11964-18
A11964-30
CAAAA11964-18EA
222.78
CAD
CAAAA11964-18
CAAAA11964-30
Methyl tiglate 98%
Methyl tiglate
Purity: 98%
CAS No.: 6622-76-0
Molecular Formula: C6H10O2
Formula Weight: 114.15
Flash Point: 39°(102°F)
Refractive Index: 1.4370
Boiling Point: 138-139°
Beilstein Reference: 1720455
Merck Reference: 14,9433
UN No.: 3272
MDL No.: MFCD00016654
Formula:
C₆H₁₀O₂ Boiling Pt: 138…139 °C Density: 0.950 Flash Pt: 39 °C(102 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00016654 CAS Number: 6622-76-0 EINECS: 229-575-8 UN: 1993 ADR: "3",III Merck Index: 14,9433 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...