N-Methylformanilide 99%

Supplier: Thermo Scientific Chemicals
Warning

A11829-30 A11829-0C A11829-0E A11829-36
CAAAAA11829-30EA 122.45 CAD
CAAAAA11829-30 CAAAA11829-0C CAAAA11829-0E CAAAA11829-36
N-Methylformanilide 99%
N-Methylformanilide

Fieser: 1,680


Purity: 99% CAS No.: 93-61-8 Molecular Formula: C8H9NO Formula Weight: 135.17 Flash Point: 126°(258°F) Refractive Index: 1.5610 Melting Point: 9-13° Boiling Point: 242-244° Beilstein Reference: 636496 MDL No.: MFCD00003283

Formula: C₈H₉NO
Boiling Pt: 242…244 °C
Melting Pt: 9…13 °C
Density: 1.095
Flash Pt: 126 °C(258 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003283
CAS Number: 93-61-8
EINECS: 202-262-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR