3-Methyl-1-phenyl-2-phospholene-1-oxide 94%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-Methyl-1-phenyl-2,3-dihydro-1H-phosphole 1-oxide
A11792-06
A11792-14
A11792-22
CAAAAA11792-06EA
130.82
CAD
CAAAAA11792-06
CAAAAA11792-14
CAAAAA11792-22
3-Methyl-1-phenyl-2-phospholene-1-oxide 94%
3-Methyl-1-phenyl-2-phospholene 1-oxide
Fieser: 1,695 2,279
Purity: 94%
CAS No.: 707-61-9
Molecular Formula: C11H13OP
Formula Weight: 192.19
Odor: Unpleasant
Melting Point: 60-65°
Boiling Point: 173-174°/0.7mm
Beilstein Reference: 609223
MDL No.: MFCD00014518
Formula:
C₁₁H₁₃OP MW: 192.2 g/mol Boiling Pt: 173…174 °C (0.7 mmHg) Melting Pt: 60…65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014518 CAS Number: 707-61-9 EINECS: 211-901-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...