3,4-Dichloro-1,2,5-thiadiazole 99%

Supplier: Thermo Scientific Chemicals
Warning

A11738-09 A11738-18 A11738-30
CAAAA11738-09EA 54.67 CAD
CAAAA11738-09 CAAAA11738-18 CAAAA11738-30
3,4-Dichloro-1,2,5-thiadiazole 99%
3,4-Dichloro-1,2,5-thiadiazole

Purity: 99% CAS No.: 5728-20-1 Molecular Formula: C2Cl2N2S Formula Weight: 155.01 Flash Point: 100°(212°F) Refractive Index: 1.5620 Boiling Point: 157-159° Beilstein Reference: 606585 UN No.: 2810 MDL No.: MFCD00010072

Formula: C₂Cl₂N₂S
Boiling Pt: 157…159 °C
Density: 1.644
Flash Pt: >100 °C(212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010072
CAS Number: 5728-20-1
EINECS: 227-232-7
UN: 2810
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR