Sodium hippurate 96%
Supplier: Thermo Scientific Chemicals
A11723-18
A11723-30
CAAAA11723-18EA
86.9
CAD
CAAAA11723-18
CAAAA11723-30
Sodium hippurate 96%
Sodium hippurate
Purity: 96%
CAS No.: 532-94-5
Molecular Formula: C6H5CONHCH2CO2Na
Formula Weight: 201.16
MDL No.: MFCD00002693
Formula:
C₉H₈NNaO₃ Storage Temperature: Ambient |
MDL Number:
MFCD00002693 CAS Number: 532-94-5 EINECS: 208-548-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...