Diethyl acetylenedicarboxylate 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Diethyl but-2-yne-1,4-dioate
A11719-14
A11719-22
A11719-36
CAAAA11719-14EA
130.35
CAD
CAAAA11719-14
CAAAA11719-22
CAAAA11719-36
Diethyl acetylenedicarboxylate 96%
Diethyl acetylenedicarboxylate
Fieser: 1,244 2,127
Purity: 96%
CAS No.: 762-21-0
Molecular Formula: CH3CH2O2CC≡CCO2CH2CH3
Formula Weight: 170.16
Flash Point: 94°(201°F)
Refractive Index: 1.4430
Melting Point: 1-2°
Boiling Point: 108-110°/11mm
Beilstein Reference: 743166
UN No.: 1760
MDL No.: MFCD00009186
Formula:
C₈H₁₀O₄ Boiling Pt: 108…110 °C/11 mm Melting Pt: 1…2 °C Density: 1.067 Flash Pt: 94 °C(201 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009186 CAS Number: 762-21-0 EINECS: 212-095-8 UN: 1760 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...