9-Fluorenylmethyl chloroformate (Fmoc chloride) 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (9H-Fluoren-9-yl)methyl carbonochloridate, 9-Fluorenylmethyl chloroformate, Fmoc-Cl, 9H-Fluoren-9-ylmethyl chloroformate, Chloroformic acid 9-fluorenylmethyl ester, FMOCCl

A11683-03 A11683-06 A11683-14
CAAAAA11683-03EA 44.4 CAD
CAAAAA11683-03 CAAAA11683-06 CAAAA11683-14
9-Fluorenylmethyl chloroformate (Fmoc chloride) 98+%
9-Fluorenylmethyl chloroformate (Fmoc chloride)

Base sensitive amino protecting group for solid-phase peptide synthesis


Purity: 98% CAS No.: 28920-43-6 Molecular Formula: C15H11ClO2 Formula Weight: 258.70 Storage Temperature: 2°C to 8°C Physical Form: Cryst powder Sensitivity: Moisture Sensitive Appearance: White to pale yellow Melting Point: 60-64° Beilstein Reference: 2279177 UN No.: 2923 MDL No.: MFCD00001138


Base sensitive amino protecting group for solid-phase peptide synthesis

Formula: C₁₅H₁₁ClO₂
MW: 258.7 g/mol
Melting Pt: 60…64 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00001138
CAS Number: 28920-43-6
EINECS: 249-313-6
UN: 2928
ADR: "6.1",II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR