3-(3,4-Dihydroxyphenyl)propionic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11638-06
A11638-14
A11638-22
CAAAA11638-06EA
102.23
CAD
CAAAA11638-06
CAAAA11638-14
CAAAA11638-22
3-(3,4-Dihydroxyphenyl)propionic acid 98+%
3-(3,4-Dihydroxyphenyl)propionic acid
Purity: 98+%
CAS No.: 1078-61-1
Molecular Formula: C9H10O4
Formula Weight: 182.18
Melting Point: 136-140°
Beilstein Reference: 2213449
Formula:
C₉H₁₀O₄ Melting Pt: 136…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002776 CAS Number: 1078-61-1 EINECS: 214-083-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...