Aluminium acetate basic hydrate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diacetoxy(hydroxy)aluminum

A11620-0E A11620-22 A11620-36
CAAAA11620-0EEA 609.88 CAD
CAAAA11620-0E CAAAA11620-22 CAAAA11620-36
Aluminium acetate basic hydrate
Aluminium acetate basic hydrate

CAS No.: 142-03-0 Molecular Formula: approx (CH3CO2)2AlOH.xH2O Physical Form: Powder Appearance: White Merck Reference: 14,344 MDL No.: MFCD00008688

Formula: C₄H₇AlO₅·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00008688
CAS Number: 142-03-0
EINECS: 205-518-2
Merck Index: 14,344

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR