Benzyl carbamate 99%
Supplier: Thermo Scientific Chemicals
A11569-30
A11569-14
A11569-22
CAAAAA11569-30EA
374.46
CAD
CAAAAA11569-30
CAAAA11569-14
CAAAA11569-22
Benzyl carbamate 99%
Benzyl carbamate
Purity: 99%
CAS No.: 621-84-1
Molecular Formula: H2NCO2CH2C6H5
Formula Weight: 151.17
Melting Point: 85-89°
Beilstein Reference: 1865635
Formula:
C₈H₉NO₂ MW: 151.17 g/mol Melting Pt: 85…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007965 CAS Number: 621-84-1 EINECS: 210-710-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...