Thiazolidine-2-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Thiazolidine-2-carboxylic acid, 2-Carboxy-1,3-thiazolidine, 1,3-Thiazolane-2-carboxylic acid

A11564-06 A11564-14
CAAAAA11564-22EA 1115.46 CAD
CAAAAA11564-22 CAAAA11564-06 CAAAA11564-14
Thiazolidine-2-carboxylic acid 97%
Thiazolidine-2-carboxylic acid

Purity: 97% CAS No.: 16310-13-7 Molecular Formula: C4H7NO2S Formula Weight: 133.17 Melting Point: ca 184° dec. Beilstein Reference: 112783

Formula: C₄H₇NO₂S
MW: 133.169 g/mol
Melting Pt: ca 184 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00068005
CAS Number: 65126-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR