5-Iodo-2'-deoxyuridine (Idoxuridine) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-5-iodopyrimidine-2,4(1H,3H)-dione, Idoxuridine, (+)-5-Iodo-2'-deoxyuridine
A11542-03
A11542-06
CAAAA11542-03EA
85.95
CAD
CAAAA11542-03
CAAAA11542-06
5-Iodo-2'-deoxyuridine (Idoxuridine) 98%
5-Iodo-2'-deoxyuridine (Idoxuridine)
Purity: 98%
CAS No.: 54-42-2
Molecular Formula: C9H11IN2O5
Formula Weight: 354.10
Odor: Odorless
Melting Point: ca 190° dec.
Beilstein Reference: 30397
Merck Reference: 14,4891
MDL No.: MFCD00134656
Formula:
C₉H₁₁IN₂O₅ MW: 354.10 g/mol Melting Pt: ca 190 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00134656 CAS Number: 54-42-2 EINECS: 200-207-8 Merck Index: 14,4891 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...